Ethyl-5-(tributylstannyl)isoxazole-3-carboxylate ≥96%
Supplier: Matrix Scientific
069467-250MG
069467-500MG
069467-1G
101940-288EA
466.15
USD
101940-288
101940-290
101940-292
Ethyl-5-(tributylstannyl)isoxazole-3-carboxylate ≥96%
Ethyl 5-(tributylstannyl)isoxazole-3-carboxylate
Hazard: Irritant
Formula:
C₁₈H₃₃NO₃Sn |
MDL Number:
MFCD07368612 CAS Number: 126085-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...