4-(Tributylstannyl)pyridazine ≥96%

Supplier: Matrix Scientific

069466-500MG 069466-1G 069466-5G
101940-282EA 538.46 USD
101940-282 101940-284 101940-286
4-(Tributylstannyl)pyridazine ≥96%
4-(Tributylstannyl)pyridazine

Hazard: Irritant

Formula: C₁₆H₃₀N₂Sn
MDL Number: MFCD07787369
CAS Number: 194865-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR