5-(Tributylstannyl)pyrimidine ≥96%

Supplier: Matrix Scientific

069464-1G 069464-5G
101940-272EA 640 USD
101940-272 101940-274
5-(Tributylstannyl)pyrimidine ≥96%
5-(Tributylstannyl)pyrimidine

Hazard: Irritant

Formula: C₁₆H₃₀N₂Sn
MW: 369.13
MDL Number: MFCD01319056
CAS Number: 144173-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR