3-Bromo-5-(4-fluorophenyl)isoxazole ≥98%

Supplier: Matrix Scientific

069460-500MG 069460-1G 069460-5G
101940-248EA 349.23 USD
101940-248 101940-250 101940-252
3-Bromo-5-(4-fluorophenyl)isoxazole ≥98%
3-Bromo-5-(4-fluorophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₅BrFNO
MDL Number: MFCD06660142
CAS Number: 903130-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR