3-Bromo-5-(hydroxymethyl)isoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: (3-Bromoisoxazol-5-yl)methanol

069459-500MG 069459-5G
101940-242EA 1978.46 USD
101940-242 101940-246
3-Bromo-5-(hydroxymethyl)isoxazole ≥95%
3-Bromo-5-(hydroxymethyl)isoxazole

Hazard: Irritant

Formula: C₄H₄BrNO₂
MW: 177.98 g/mol
MDL Number: MFCD04035586
CAS Number: 25742-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR