Ethyl-4-oxazolecarboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl oxazole-4-carboxylate

069457-1G 069457-5G
101940-232EA 116.92 USD
101940-232 101940-234
Ethyl-4-oxazolecarboxylate ≥98%
Ethyl 4-oxazolecarboxylate

Hazard: Irritant

Formula: C₆H₇NO₃
MW: 141.1300 g/mol
MDL Number: MFCD04114940
CAS Number: 23012-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR