1-Tritylimidazole-4-carbaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Trityl-1H-imidazole-4-carboxaldehyde, 1-Trityl-1H-imidazole-4-carbaldehyde, 1-Tritylimidazole-4-carboxaldehyde

069456-5G 069456-25G
101940-226EA 60 USD
101940-226 101940-228
1-Tritylimidazole-4-carbaldehyde ≥98%
1-Tritylimidazole-4-carbaldehyde

Hazard: Irritant

Formula: C₂₃H₁₈N₂O
MW: 338.41 g/mol
Melting Pt: 187…192 °C (dec.)
MDL Number: MFCD02179554
CAS Number: 33016-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR