5-Thiazolemethanol

Supplier: Matrix Scientific

Synonyms: Thiazol-5-ylmethanol, 5-(Hydroxymethyl)thiazole, Thiazole-5-methanol

069455-5G 069455-25G 069455-100G
101940-218EA 175.38 USD
101940-218 101940-220 101940-222
5-Thiazolemethanol
5-Thiazolemethanol

Hazard: Irritant

Formula: C₄H₅NOS
MW: 115.15 g/mol
MDL Number: MFCD04115732
CAS Number: 38585-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR