4-(Hydroxymethyl)thiazole ≥97%

Supplier: Matrix Scientific

069454-1G 069454-5G
101940-214EA 110.77 USD
101940-214 101940-216
4-(Hydroxymethyl)thiazole ≥97%
4-(Hydroxymethyl)thiazole

Hazard: Irritant

Formula: C₄H₅NOS
Boiling Pt: 123…125 °C (15 Torr)
Melting Pt: 112…114 °C
MDL Number: MFCD04115728
CAS Number: 7036-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR