3-Carboxyphenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

069453-1G 069453-5G 069453-25G
101940-208EA 56.92 USD
101940-208 101940-210 102551-752
3-Carboxyphenylboronic acid pinacol ester ≥98%
3-Carboxyphenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₁₇BO₄
MW: 248.083 g/mol
MDL Number: MFCD03411930
CAS Number: 269409-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR