Methyl-2-bromothiazole-5-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 2-bromothiazole-5-carboxylate

069451-5G 069451-25G
101940-196EA 93.85 USD
101940-196 101940-198
Methyl-2-bromothiazole-5-carboxylate ≥98%
Methyl-2-bromothiazole-5-carboxylate

Hazard: Irritant

Formula: C₅H₄BrNO₂S
MW: 222.05 g/mol
MDL Number: MFCD03788565
CAS Number: 54045-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR