Methyl-2-aminothiazole-5-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 2-aminothiazole-5-carboxylate, 2-Aminothiazole-5-carboxylic acid methyll ester

069449-5G 069449-25G
101940-184EA 73.85 USD
101940-184 101940-186
Methyl-2-aminothiazole-5-carboxylate ≥98%
Methyl-2-aminothiazole-5-carboxylate

Hazard: Irritant

Formula: C₅H₆N₂O₂S
MDL Number: MFCD03788562
CAS Number: 6633-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR