Methyl-2-amino-5-bromothiazole-4-carboxylate ≥96%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-5-bromothiazole-4-carboxylate

069447-5G 069447-25G
101940-172EA 143.08 USD
101940-172 101940-174
Methyl-2-amino-5-bromothiazole-4-carboxylate ≥96%
Methyl-2-amino-5-bromothiazole-4-carboxylate

Hazard: Irritant

Formula: C₅H₅BrN₂O₂S
MW: 237.08 g/mol
MDL Number: MFCD00137990
CAS Number: 850429-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR