Ethyl thiazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

069444-1G 069444-5G
101940-154EA 307.69 USD
101940-154 101940-156
Ethyl thiazole-4-carboxylate ≥97%
Ethyl thiazole-4-carboxylate

Hazard: Irritant

Formula: C₆H₇NO₂S
MDL Number: MFCD03788566
CAS Number: 14527-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR