4-Bromo-1-tritylimidazole ≥98%

Supplier: Matrix Scientific

069443-1G 069443-5G
101940-150EA 167.69 USD
101940-150 101940-152
4-Bromo-1-tritylimidazole ≥98%
4-Bromo-1-tritylimidazole

Hazard: Irritant

Formula: C₂₂H₁₇BrN₂
MDL Number: MFCD00185657
CAS Number: 87941-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR