2-Bromo-5-hydroxymethylthiazole ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromothiazole-5-methanol

069442-500MG 069442-1G 069442-5G
101940-144EA 233.85 USD
101940-144 101940-146 101940-148
2-Bromo-5-hydroxymethylthiazole ≥97%
2-Bromo-5-hydroxymethylthiazole

Hazard: Irritant

Formula: C₄H₄BrNOS
MW: 194.06 g/mol
MDL Number: MFCD04115733
CAS Number: 687636-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR