Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-amino-1H-pyrrole-2-carboxylate hydrochloride

069435-1G 069435-5G
101940-104EA 83.08 USD
101940-104 101940-106
Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride ≥95%
Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride

Hazard: Irritant

Formula: C₇H₁₁ClN₂O₂
MW: 190.6274 g/mol
Melting Pt: 198…204 °C
MDL Number: MFCD04038917
CAS Number: 252932-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR