3-Bromo-5-(chloromethyl)isoxazole ≥98%

Supplier: Matrix Scientific

069434-500MG 069434-1G 069434-5G
101940-098EA 230.77 USD
101940-098 101940-100 101940-102
3-Bromo-5-(chloromethyl)isoxazole ≥98%
3-Bromo-5-(chloromethyl)isoxazole

Hazard: Irritant/Refrigerate

Formula: C₄H₃BrClNO
MDL Number: MFCD06660137
CAS Number: 124498-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR