7-Methylindole-3-carboxyaldehyde ≥96%

Supplier: Matrix Scientific

Synonyms: 7-Methyl-1H-indole-3-carbaldehyde

069433-500MG 069433-1G 069433-5G
101940-092EA 87.69 USD
101940-092 101940-094 101940-096
7-Methylindole-3-carboxyaldehyde ≥96%
7-Methylindole-3-carboxyaldehyde

Hazard: Irritant

Formula: C₁₀H₉NO
MDL Number: MFCD00047170
CAS Number: 4771-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR