Methyl (2-chloromethyl)oxazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(chloromethyl)oxazole-4-carboxylate

069431-500MG 069431-1G 069431-5G
101940-080EA 233.85 USD
101940-080 101940-082 101940-084
Methyl (2-chloromethyl)oxazole-4-carboxylate ≥97%
Methyl (2-chloromethyl)oxazole-4-carboxylate

Hazard: Irritant

Formula: C₆H₆ClNO₃
MW: 175.57 g/mol
MDL Number: MFCD06660133
CAS Number: 208465-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR