Diethyl-2-isopropylthiazole-4-methylphosphonate ≥98%

Supplier: Matrix Scientific

069430-500MG 069430-1G
101940-076EA 1106.15 USD
101940-076 101940-078
Diethyl-2-isopropylthiazole-4-methylphosphonate ≥98%
Diethyl-2-isopropylthiazole-4-methylphosphonate

Hazard: Irritant

Formula: C₁₁H₂₀NO₃PS
MDL Number: MFCD06660132
CAS Number: 675201-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR