o-Carborane ≥99%

Supplier: Matrix Scientific

069428-1G 069428-5G 069428-25G
101940-068EA 169.23 USD
101940-068 101940-070 101940-072
o-Carborane ≥99%
o-Carborane

TCA listed: T
Hazard: Flammable Solid

Formula: C₂H₁₂B₁₀
MW: 144.23 g/mol
Melting Pt: 285…287 °C
Density: 0.95
MDL Number: MFCD00012169
CAS Number: 16872-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR