1-Benzyl-2-(chloromethyl)-1H-imidazole hydrochloride ≥97%
Supplier: Matrix Scientific
069427-1G
069427-5G
069427-25G
101940-062EA
233.85
USD
101940-062
101940-064
101940-066
1-Benzyl-2-(chloromethyl)-1H-imidazole hydrochloride ≥97%
1-Benzyl-2-(chloromethyl)-1H-imidazole hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₂Cl₂N₂ |
MDL Number:
MFCD00035210 CAS Number: 19276-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...