4-(Chloromethyl)-1H-imidazole hydrochloride ≥96%

Supplier: Matrix Scientific

069426-500MG 069426-1G 069426-5G
101940-056EA 84.62 USD
101940-056 101940-058 101940-060
4-(Chloromethyl)-1H-imidazole hydrochloride ≥96%
4-(Chloromethyl)-1H-imidazole hydrochloride

Hazard: Irritant

Formula: C₄H₅ClN₂·ClH
MW: 153.01 g/mol
MDL Number: MFCD01721661
CAS Number: 38585-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR