2-Bromo-4-methylthiazole ≥95%

Supplier: Matrix Scientific

069425-1G 069425-5G 069425-25G
101940-050EA 84.62 USD
101940-050 101940-052 101940-054
2-Bromo-4-methylthiazole ≥95%
2-Bromo-4-methylthiazole

Hazard: Irritant

Formula: C₄H₄BrNS
MW: 178.05 g/mol
MDL Number: MFCD06660130
CAS Number: 7238-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR