4-Chloromethyl-2-methylthiazole hydrochloride ≥97%

Supplier: Matrix Scientific

069424-5G 069424-25G
101940-046EA 349.23 USD
101940-046 101940-048
4-Chloromethyl-2-methylthiazole hydrochloride ≥97%
4-Chloromethyl-2-methylthiazole hydrochloride

Hazard: Irritant

Formula: C₅H₇Cl₂NS
MW: 184.078 g/mol
MDL Number: MFCD00067725
CAS Number: 77470-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR