4-(Chloromethyl)-2-isopropylthiazole ≥98%

Supplier: Matrix Scientific

069423-500MG 069423-1G 069423-5G
101940-040EA 349.23 USD
101940-040 101940-042 101940-044
4-(Chloromethyl)-2-isopropylthiazole ≥98%
4-(Chloromethyl)-2-isopropylthiazole

Hazard: Irritant

Formula: C₇H₁₀ClNS
MW: 175.68 g/mol
MDL Number: MFCD06660126
CAS Number: 40516-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR