4-Formyl-2-methylthiazole ≥96%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-1,3-thiazole-4-carboxaldehyde, 2-Methylthiazole-4-carbaldehyde

069422-1G 069422-5G
101940-036EA 223.08 USD
101940-036 101940-038
4-Formyl-2-methylthiazole ≥96%
4-Formyl-2-methylthiazole

Hazard: Irritant

Formula: C₅H₅NOS
MW: 127.1643 g/mol
MDL Number: MFCD06655197
CAS Number: 20949-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR