4-Bromo-2-formylthiazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1,3-thiazole-2-carboxaldehyde

069420-500MG 069420-1G 069420-5G
101940-022EA 116.92 USD
101940-022 101940-024 101940-026
4-Bromo-2-formylthiazole ≥97%
4-Bromo-2-formylthiazole

Hazard: Irritant

Formula: C₄H₂BrNOS
MW: 192.0338 g/mol
MDL Number: MFCD06660122
CAS Number: 167366-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR