(1-Trityl-1H-imidazol-4-yl)methanol ≥99%

Supplier: Matrix Scientific

069419-1G 069419-5G 069419-25G
101940-016EA 524.62 USD
101940-016 101940-018 101940-020
(1-Trityl-1H-imidazol-4-yl)methanol ≥99%
(1-Trityl-1H-imidazol-4-yl)methanol

Hazard: Irritant

Formula: C₂₃H₂₀N₂O
MW: 340.4177 g/mol
MDL Number: MFCD02179553
CAS Number: 33769-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR