(1H-Imidazol-4-yl)methanol hydrochloride ≥97%
Supplier: Matrix Scientific
069418-1G
069418-5G
069418-25G
101940-010EA
96.92
USD
101940-010
101940-012
101940-014
(1H-Imidazol-4-yl)methanol hydrochloride ≥97%
(1H-Imidazol-4-yl)methanol hydrochloride
Hazard: Irritant
Formula:
C₄H₇ClN₂O |
MDL Number:
MFCD00012697 CAS Number: 32673-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...