3-(tert-Butoxycarbonyl)phenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(tert-Butoxycarbonyl)benzeneboronic acid, 3-tert-Butoxycarbonylphenylboronic acid

069417-1G 069417-5G
101940-004EA 27.69 USD
101940-004 101940-006
3-(tert-Butoxycarbonyl)phenylboronic acid ≥98%
3-(tert-Butoxycarbonyl)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₁₁H₁₅BO₄
MW: 222.05 g/mol
Melting Pt: 96…102 °C
MDL Number: MFCD01630855
CAS Number: 220210-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR