tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Supplier: Matrix Scientific

Synonyms: N-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-(Boc-amino)phenylboronic acid Pinacol ester, tert-Butyl [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate, 4-[(tert-Butoxycarbonyl)amino]phenylboronic acid Pinacol ester, 4-Boc-aminophenylboronic acid, pinacol ester, N-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamic acid tert-butyl ester, 4-(Boc-amino)benzeneboronic acid pinacol ester

069416-1G 069416-5G 069416-25G
101939-998EA 220 USD
101939-998 101940-000 101940-002
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Hazard: Irritant

Formula: C₁₇H₂₆BNO₄
MDL Number: MFCD02179439
CAS Number: 330793-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR