2-Methylthiazole-5-methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Methylthiazol-5-yl)methanol

069414-1G 069414-5G
101939-990EA 930.77 USD
101939-990 101939-992
2-Methylthiazole-5-methanol ≥95%
2-Methylthiazole-5-methanol

Hazard: Irritant

Formula: C₅H₇NOS
MW: 129.18 g/mol
MDL Number: MFCD12912988
CAS Number: 56012-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR