Triphenylphosphine imine sulfate ≥97%

Supplier: Matrix Scientific

069410-1G 069410-5G
101939-978EA 246.15 USD
101939-978 101939-980
Triphenylphosphine imine sulfate ≥97%
Triphenylphosphine imine sulfate

Hazard: Irritant

Formula: C₁₈H₁₈NO₄PS
MW: 375.39 g/mol
MDL Number: MFCD22422287
CAS Number: 117672-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR