2,5-Diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2,5-diazaspiro[3.4]octane-2-carboxylate

069407-250MG 069407-500MG 069407-1G
101939-966EA 1512.31 USD
101939-966 101939-968 101939-970
2,5-Diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester ≥95%
2,5-Diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD11501185
CAS Number: 1086398-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR