2-Chloro-4-hydrazinopyrimidine ≥95%

Supplier: Matrix Scientific

065800-500MG 065800-1G
101930-382EA 141.54 USD
101930-382 101930-384
2-Chloro-4-hydrazinopyrimidine ≥95%
2-Chloro-4-hydrazinopyrimidine

Hazard: Irritant

Formula: C₄H₅ClN₄
MW: 144.56 g/mol
MDL Number: MFCD06658201
CAS Number: 52476-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR