2-Chloronicotinoyl chloride
Supplier: Matrix Scientific
065589-5G
065589-25G
101929-980EA
43.08
USD
101929-980
101929-982
2-Chloronicotinoyl chloride
2-Chloronicotinoyl chloride
Hazard: Corrosive/Irritant
Formula:
C₆H₃Cl₂NO MW: 176.00 g/mol Melting Pt: 38…42 °C |
MDL Number:
MFCD00051677 CAS Number: 49609-84-9 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...