Benzofuran-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

065543-5G 065543-25G
101929-886EA 52.31 USD
101929-886 101929-888
Benzofuran-2-carboxylic acid ≥95%
Benzofuran-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆O₃
Boiling Pt: 310-315°C
Melting Pt: 192-196°C
MDL Number: MFCD00005848
CAS Number: 496-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR