3-(4-Methylpiperazin-1-yl)propan-1-ol dihydrochloride ≥95%

Supplier: Matrix Scientific

060693-5G 060693-25G
101920-498EA 106.15 USD
101920-498 101920-500
3-(4-Methylpiperazin-1-yl)propan-1-ol dihydrochloride ≥95%
3-(4-Methylpiperazin-1-yl)propan-1-ol dihydrochloride

Hazard: Irritant

Formula: C₈H₂₀Cl₂N₂O
MDL Number: MFCD11841273
CAS Number: 82980-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR