6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Supplier: Matrix Scientific
060176-5G
060176-25G
101919-554EA
56.92
USD
101919-554
101919-556
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₆ClNO₂ MW: 229.71 g/mol Melting Pt: 263 °C |
MDL Number:
MFCD00012744 CAS Number: 2328-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...