1-Boc-3-(methylaminomethyl)azetidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl 3-[(methylamino)methyl]azetidine-1-carboxylate, 3-[(Methylamino)methyl]azetidine, N1-BOC protected, 1-Boc-3-((Methylamino)methyl)azetidine

059929-1G 059929-250MG
101919-470EA 633.85 USD
101919-470 102551-556
1-Boc-3-(methylaminomethyl)azetidine ≥95%
1-Boc-3-(methylaminomethyl)azetidine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.282 g/mol
MDL Number: MFCD08061965
CAS Number: 1049730-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR