tert-Butyl-3-iodoazetidine-1-carboxylate ≥96%

Supplier: Matrix Scientific

Synonyms: Tert-butyl 3-iodoazetidine-1-carboxylate, 1-BOC-3-iodoazetidine, tert-Butyl 3-iodoazetidine-1-carboxylate, N-Boc-3-iodoazetidine

059924-5G 059924-25G
101919-458EA 64.62 USD
101919-458 101919-460
tert-Butyl-3-iodoazetidine-1-carboxylate ≥96%
tert-Butyl-3-iodoazetidine-1-carboxylate

Hazard: Irritant

Formula: C₈H₁₄INO₂
MW: 283.11 g/mol
MDL Number: MFCD09752821
CAS Number: 254454-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR