Methyl-1-boc-azetidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

059923-1G 059923-5G 059923-25G
101919-456EA 15.38 USD
101919-456 102833-294 102833-296
Methyl-1-boc-azetidine-3-carboxylate ≥95%
Methyl-1-boc-azetidine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MDL Number: MFCD06656775
CAS Number: 610791-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR