Ethyl-3-amino-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-amino-1H-pyrrole-2-carboxylate

059891-250MG 059891-1G
101919-418EA 235.38 USD
101919-418 101919-420
Ethyl-3-amino-1H-pyrrole-2-carboxylate ≥95%
Ethyl-3-amino-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MW: 154.1665 g/mol
MDL Number: MFCD06797655
CAS Number: 252932-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR