4-Chloro-1H-pyrazolo[3,4-d]pyrimidine ≥97%

Supplier: Matrix Scientific

059889-1G 059889-5G
101919-414EA 93.85 USD
101919-414 102833-292
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine ≥97%
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine

Hazard: Irritant

Formula: C₅H₃ClN₄
MDL Number: MFCD03030404
CAS Number: 5399-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR