7-Bromo-4-chlorothieno[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

059885-250MG 059885-1G
101919-408EA 218.46 USD
101919-408 101919-410
7-Bromo-4-chlorothieno[3,2-d]pyrimidine ≥95%
7-Bromo-4-chlorothieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆H₂BrClN₂S
MW: 249.5200 g/mol
MDL Number: MFCD08273917
CAS Number: 31169-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR