7-Bromo-2,4-dichlorothieno[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

059882-250MG 059882-1G
101919-404EA 235.38 USD
101919-404 101919-406
7-Bromo-2,4-dichlorothieno[3,2-d]pyrimidine ≥95%
7-Bromo-2,4-dichlorothieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆HBrCl₂N₂S
MDL Number: MFCD11869289
CAS Number: 41102-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR