2,4-Dichlorothieno[3,2-d]pyrimidine ≥97%

Supplier: Matrix Scientific

059881-1G 059881-5G
101919-402EA 15.38 USD
101919-402 102833-290
2,4-Dichlorothieno[3,2-d]pyrimidine ≥97%
2,4-Dichlorothieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆H₂Cl₂N₂S
MW: 205.0645 g/mol
MDL Number: MFCD08448158
CAS Number: 16234-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR