7-Bromo-4-chloro-5H-pyrrolo[3,2-d]pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

059879-250MG 059879-1G
101919-394EA 347.69 USD
101919-394 101919-396
7-Bromo-4-chloro-5H-pyrrolo[3,2-d]pyrimidin-2-amine ≥95%
7-Bromo-4-chloro-5H-pyrrolo[3,2-d]pyrimidin-2-amine

Hazard: Irritant

Formula: C₆H₄BrClN₄
MDL Number: MFCD19105254
CAS Number: 1311275-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR